Check Order Status
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEJ875008
also known as GEEJ/8750/08
500 mL
Amber glass bottle
Catalog number
GEEJ875008
also known as GEEJ/8750/08
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS
2052-49-5
Color
Colorless
Physical Form
Liquid
Quantity
500 mL
Molecular Formula
C16H37NO
View more
Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis