Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Usp, Fisher Chemical
Have Questions?
Catalog NumberQuantity
GEEJ733015
also known as GEEJ/7330/15
1 L
Catalog number GEEJ733015
also known as GEEJ/7330/15
Price (SGD)
106.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
106.00
EA
Add to cart
Specifications
ColorColorless
CAS7761-88-8
Molecular FormulaAgNO3
MDL NumberMFCD00003414
Synonymsilver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
View more
Rready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP