Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
GEEJ435015
also known as GEEJ/4350/15
1 L
Catalog number GEEJ435015
also known as GEEJ/4350/15
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
pH2
ColorColorless
CAS7647-01-0
Boiling Point100°C
PackagingHDPE plastic bottle
View more
ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol