Check Order Status
Hydrochloric Acid Solution 2M (2N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 2M (2N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEJ431515
also known as GEEJ/4315/15
1 L
HDPE plastic bottle
Catalog number
GEEJ431515
also known as GEEJ/4315/15
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Color
Colorless
CAS
7647-01-0
Boiling Point
100°C
Packaging
HDPE plastic bottle
Quantity
1 L
View more