Check Order Status
Potassium Chloride Solution 4M, Pure, Electrode Filling Solution with Silver Chloride, Fisher Chemical
Element Search
Structure Search
Potassium Chloride Solution 4M, Pure, Electrode Filling Solution with Silver Chloride, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
GEEJ298405
also known as GEEJ/2984/05
100 mL
Amber glass bottle
Catalog number
GEEJ298405
also known as GEEJ/2984/05
Price (SGD)
-
Quantity:
100 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS
7447-40-7
Color
Colorless
Physical Form
Liquid
Quantity
100 mL
Molecular Formula
ClK
View more