Check Order Status
Hexanes (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Hexanes (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label.
Have Questions?
Catalog Number
Quantity
Packaging
GEEH292-20
20 L
Steel Pail
Catalog number
GEEH292-20
Price (SGD)
347.00
EA
Add to cart
Quantity:
20 L
Packaging:
Steel Pail
Request bulk or custom format
Price (SGD)
347.00
EA
Add to cart
Specifications
Chemical Name or Material
Hexanes
Cleanup Symbol
Solvent
CAS
92112-69-1
ChemAlert Storage Symbol
Red
Color
Colorless
View more