Check Order Status
Hydrogen Peroxide 12% (w/v) (40 Volumes), Pure, Fisher Chemical
Element Search
Structure Search
Hydrogen Peroxide 12% (w/v) (40 Volumes), Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
GEEH165017
also known as GEEH/1650/17
2.5 L
Catalog number
GEEH165017
also known as GEEH/1650/17
Price (SGD)
114.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
114.00
EA
Add to cart
Specifications
Color
Colorless
CAS
7722-84-1
Boiling Point
100°C
Melting Point
0°C
Molecular Formula
H2O2
View more