Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
ClH, CAS Number-7647-01-0, muriatic acid, spirits of salt, hydrogen chloride hcl, hydrogen chloride, hydrochloric acid, chlorwasserstoff, chloorwaterstof, chlorohydric acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
HDPE plastic bottle
Catalog number GEEH1200PC17
also known as GEEH/1200/PC17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
pH1
ColorColorless
CAS7647-01-0
Melting Point-35°C
Boiling Point57°C
View more
ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, chlorwasserstoff, chloorwaterstof, muriatic acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol