Check Order Status
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
Element Search
Structure Search
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C
Have Questions?
Catalog Number
Quantity
Packaging
GEEH035517
also known as GEEH/0355/17
2.5 L
Amber Glass Bottle
Catalog number
GEEH035517
also known as GEEH/0355/17
Price (SGD)
85.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (SGD)
85.00
EA
Add to cart
Specifications
Melting Point
-95°C
CAS
92112-69-1
Color
Colorless
Boiling Point
69°C
Quantity
2.5 L
View more
C6H14, CAS Number-92112-69-1, hexanes, dipropyl, gettysolve-b, skellysolve b, esani, n-hexane, heksan, hexyl hydride, hexanen, hexan, 5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Metal can, Liquid, 8058, CCCCCC, 2.97, 160mbar at 20 deg.C