Check Order Status
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Element Search
Structure Search
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C
Have Questions?
Catalog Number
Quantity
Packaging
GEED185217
also known as GEED/1852/17
2.5 L
Amber Glass Bottle
Catalog number
GEED185217
also known as GEED/1852/17
Price (SGD)
49.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (SGD)
49.00
BOT
Add to cart
Specifications
Melting Point
-97°C
CAS
75-09-2
Color
Colorless
Boiling Point
39°C
Quantity
2.5 L
View more
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C