Check Order Status
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™
DMAC processed to high purity for accurate and repeatable determination of trace levels of residual solvents.
Have Questions?
Catalog Number
Quantity
Packaging
GEED160-1
1 L
Glass Bottle
Catalog number
GEED160-1
Price (SGD)
320.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
320.00
EA
Add to cart
Specifications
Chemical Name or Material
DMAC, Dimethylacetamide
Absorbance
0.300 AU Maximum (275nm), 0.080 AU Maximum (300nm), 0.005 AU Maximum (350nm)
CAS
127-19-5
Color
Colorless
Recommended Storage
Room temperature, packaged under inert atmosphere
View more
Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
This product is UV Absorbance Tested to ensure low organic contamination
Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC