Check Order Status
Cyclohexane (HPLC), Fisher Chemical™
Element Search
Structure Search
Cyclohexane (HPLC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEC893617
also known as GEEC/8936/17
2.5 L
Amber Glass Bottle
Catalog number
GEEC893617
also known as GEEC/8936/17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Melting Point
6.5°C
CAS
110-82-7
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more