▸
Organic compounds
▸
Organic oxides
▸
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents
Have Questions?
Change view
Quantity:
500 mL
25 mL
Packaging:
View all
HDPE Bottle
Catalog number
GEEBP531-500
Price (SGD)
67.00
Each
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (SGD)
67.00
Each
Add to cart
Specifications
Chemical Name or Material
Formaldehyde
Name Note
37% by Weight
CAS
50-00-0
ChemAlert Storage Symbol
Red
Health Hazard 2
DANGER!
View more
Molecular Biology-grade Formaldehyde is used for denaturing RNA.