Check Order Status
Potassium Chloride (White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Potassium Chloride (White Crystals), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
GEEBP366-500
500 g
Poly Bottle
Catalog number
GEEBP366-500
Price (SGD)
92.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
92.00
EA
Add to cart
Specifications
Boiling Point
1420°C
Chemical Name or Material
Potassium Chloride
Melting Point
770°C
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
View more
Potassium Chloride is used in buffers for molecular biology and cell culture applications.