▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Have Questions?
Change view
Quantity:
1 L
200 mL
Packaging:
View all
Nalgene Poly Bottle
Catalog number
GEEBP2436-1
Price (SGD)
156.00
Each
Add to cart
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Price (SGD)
156.00
Each
Add to cart
Specifications
Absorbance
0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Chemical Name or Material
Sodium Dodecyl Sulfate
Color
Colorless
Name Note
10% Solution
CAS
151-21-3
View more
DNase, RNase, and protease-free premixed solution containing 10% SDS for protein extraction and DNA extraction procedures
Reduce preparation time by using this premade SDS solution
SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
Intended for research use only; not for human or animal diagnostic purposes