Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Buffers and Standards
▸
Life Science Buffers
▸
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change view
Quantity:
500 g
Packaging:
Poly Bottle
Catalog number
GEEBP152-500
Price (SGD)
114.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
114.00
Each
Add to cart
Specifications
Absorbance
0.05 AU max. at 280nm
Chemical Name or Material
Tris Base
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Melting Point
168.5°C
View more
DNase-, RNase- and Protease-Free.
Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.