Check Order Status
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
GEEB525108
also known as GEEB/5251/08
500 mL
Catalog number
GEEB525108
also known as GEEB/5251/08
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
pH
7
Melting Point
25°C
CAS
75-65-0
Boiling Point
83°C
Quantity
500 mL
View more