Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Thermo Scientific Chemicals
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
GEEA509-P500500 mLHDPE Bottle
Catalog number GEEA509-P500
Price (SGD)
175.00
EA
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (SGD)
175.00
EA
Add to cart
Specifications
Chemical Name or MaterialNitric Acid
Shelf Life2 years
pH1.0
Cleanup SymbolAcid
ColorColorless Yellow
View more
Nitric Acid (TraceMetal™ Grade)
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured,purified, and packaged in a Class 1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Two year shelf life from time of manufacture - available on COA. 45 mm bottle neck size