Check Order Status
Nitric Acid 67-70%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Nitric Acid 67-70%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, acidum nitricum, acide nitrique, aqua fortis, azotic acid, salpetersaeure, hydrogen nitrate, nitryl hydroxide, rfna, nital, nitrous fumes, 1L, CHEBI:48107, Colorless Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, Liquid, 944 | CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Catalog Number
Quantity
GEEA509-P212
2.5 L
Catalog number
GEEA509-P212
Price (SGD)
368.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
368.00
EA
Add to cart
Specifications
pH
1
Color
Yellow
CAS
7697-37-2
Melting Point
-41°C
Vapor Pressure
0.94kPa (20°C)
View more
HNO3, CAS Number-7697-37-2, 500mL, Yellow, 63.02g/mol, -41 deg.C, 11349, Liquid, 0.94kPa (20 deg.C)