Check Order Status
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
Structure Search
Thermo Scientific Chemicals
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEA508-P212
2.5 L
HDPE Bottle
Catalog number
GEEA508-P212
Price (SGD)
360.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE Bottle
Request bulk or custom format
Price (SGD)
360.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid
Shelf Life
3 years
pH
1
Cleanup Symbol
Acid
Color
Colorless
View more