Check Order Status
Ammonia Solution, 25%, Certified AR for Analysis, d=0.91, Fisher Chemical™
Element Search
Structure Search
Ammonia Solution, 25%, Certified AR for Analysis, d=0.91, Fisher Chemical™
H5NO, CAS Number-1336-21-6, 2.5L, 38 deg.C, Colorless, -57 deg.C, 66650, Plastic Coated Glass bottle, 12, Liquid, 0.59, 500 hPa at 20 deg.C
Have Questions?
Catalog Number
Quantity
GEEA3360PB15
also known as GEEA/3360/PB15
1 L
Catalog number
GEEA3360PB15
also known as GEEA/3360/PB15
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
pH
12
Color
Colorless
Boiling Point
38°C
CAS
1336-21-6
Melting Point
-57°C
View more
H5NO, CAS Number-1336-21-6, 2.5L, 38 deg.C, Colorless, -57 deg.C, 66650, Plastic Coated Glass bottle, 12, Liquid, 0.59, 500 hPa at 20 deg.C