Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Thermo Scientific Chemicals
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL
Have Questions?
Catalog NumberQuantityPackaging
GEEA116-5050 mLAmber Glass
Catalog number GEEA116-50
Price (SGD)
147.00
EA
Add to cart
Quantity:
50 mL
Packaging:
Amber Glass
Request bulk or custom format
Price (SGD)
147.00
EA
Add to cart
Specifications
Absorbance0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm
Chemical Name or MaterialTrifluoroacetic Acid
pH0.45
CAS76-05-1
ChemAlert Storage SymbolWhite
View more
Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL
  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent