Check Order Status
Ammonium Formate, Optima™ LC/MS Grade, Fisher Chemical™
Element Search
Structure Search
Thermo Scientific Chemicals
Ammonium Formate, Optima™ LC/MS Grade, Fisher Chemical™
CH5NO2, CAS Number-540-69-2, 50g, White, 63.06g/mol, 115 deg.C, 13103, 6.0 to 7.0, Solid
Have Questions?
Catalog number
GEEA115-50
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Melting Point
115°C
CAS
540-69-2
Color
White
pH
6.0 to 7.0
Physical Form
Solid
View more
CH5NO2, CAS Number-540-69-2, 50g, White, 63.06g/mol, 115 deg.C, 13103, 6.0 to 7.0, Solid