Check Order Status
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Element Search
Structure Search
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, CHEBI:15347, Colorless, 58.08g/mol, CSCPPACGZOOCGX-UHFFFAOYSA-N, propan-2-one, -95 deg.C, 8765, Amber glass bottle, Liquid, 180, CC(-O)C, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C | CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
GEEA060021
also known as GEEA/0600/21
5 L
Metal Can
Catalog number
GEEA060021
also known as GEEA/0600/21
Price (SGD)
92.00
EA
Add to cart
Quantity:
5 L
Packaging:
Metal Can
Request bulk or custom format
Price (SGD)
92.00
EA
Add to cart
Chemical Identifiers
CAS
67-64-1
Specifications
Packaging
Metal Can
CAS
67-64-1
Vapor Pressure
247mbar at 20°C
Viscosity
0.32 mPaS at 20°C
Vapor Density
2
View more
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C