Isotemp™ General Purpose Deluxe Water Baths
Actual product may vary
Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Choose general purpose water baths, designed to provide consistent temperature for reliable results with easy-to-use operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
FTNFSGPD1010 LIsotemp GPD 10
FTNFSGPD022 LIsotemp GPD 02
FKS153658772 LIsotemp GPD 2S
FTNFSGPD055 LIsotemp GPD 05
FTNFSGPD15D15 L (5 L/10 L)Isotemp GPD 15D
FTNFSGPD2020 LIsotemp GPD 20
FTNFSGPD2828 LIsotemp GPD 28
Catalog number FTNFSGPD10
Price (SGD)
1,706.00
Each
Capacity (Metric):
10 L
Model:
Isotemp GPD 10
Price (SGD)
1,706.00
Each
Ideal for industrial, clinical pharmaceutical or biomedical applications, Fisherbrand™ Isotemp™ general purpose deluxe water baths provide powerful technology in a compact design. Available in a variety of sizes to meet a wide range of application requirements, the Isotemp water baths are designed to provide consistent temperature for reliable results with easy-to-use operation.
  • Smaller footprint, compared to previous models
  • Advanced temperature controller is designed for extended functionality 
  • New audible alarms
  • Conveniently save commonly used settings with temperature presets
  • Simple to operate and monitor with icon-based graphical display
  • Enhanced hinge design supports open lid configuration and easy lid removal
  • Help prevent bath damage and overheating with low-fluid protection
  • Compatable with Thermal beads (sold separately)
Specifications
Chamber MaterialStainless Steel
Controller TypeDigital
Description10L Deluxe General Purpose Water Bath
Capacity (Metric)10 L
Certifications/ComplianceUL, CE, FDA Class I
Temperature RangeAmbient to 100°C
DisplayMonochrome LCD
Heating Capacity800 w
IncludesPolycarbonate Gable Cover, Diffuser Tray
MaterialEpoxy Powder-coated Steel
ModelIsotemp GPD 10
Temperature Presets3
Temperature Stability±0.2°C
Temperature Uniformity±0.2°C
Dimensions (D x W x H) Interior11.9 x 13 x 5.99 in. (301 x 330 x 150 mm)
Dimensions (L x W x H)15.5 x 15.1 x 9.2 in. (393 x 383 x 233 mm)
Amperage6.0-7.0 / 2.8-3.4 (120V/230V)
Unit SizeEach