Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Actual product may vary
Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Isotemp™ General Purpose Deluxe Water Baths
Choose general purpose water baths, designed to provide consistent temperature for reliable results with easy-to-use operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
FTNFSGPD055 LIsotemp GPD 05
FTNFSGPD022 LIsotemp GPD 02
FKS153658772 LIsotemp GPD 2S
FTNFSGPD1010 LIsotemp GPD 10
FTNFSGPD15D15 L (5 L/10 L)Isotemp GPD 15D
FTNFSGPD2020 LIsotemp GPD 20
FTNFSGPD2828 LIsotemp GPD 28
Catalog number FTNFSGPD05
Price (SGD)
1,216.00
Special offer
Ends: 31-Dec-2025
1,368.00
Save 152.00 (11%)
EA
Add to cart
Capacity (Metric):
5 L
Model:
Isotemp GPD 05
Price (SGD)
1,216.00
Special offer
Ends: 31-Dec-2025
1,368.00
Save 152.00 (11%)
EA
Add to cart
Ideal for industrial, clinical pharmaceutical or biomedical applications, Fisherbrand™ Isotemp™ general purpose deluxe water baths provide powerful technology in a compact design. Available in a variety of sizes to meet a wide range of application requirements, the Isotemp water baths are designed to provide consistent temperature for reliable results with easy-to-use operation.
  • Smaller footprint, compared to previous models
  • Advanced temperature controller is designed for extended functionality 
  • New audible alarms
  • Conveniently save commonly used settings with temperature presets
  • Simple to operate and monitor with icon-based graphical display
  • Enhanced hinge design supports open lid configuration and easy lid removal
  • Help prevent bath damage and overheating with low-fluid protection
  • Compatable with Thermal beads (sold separately)
Specifications
Chamber MaterialStainless Steel
Controller TypeDigital
Description5L Deluxe General Purpose Water Bath
Capacity (Metric)5 L
Certifications/ComplianceUL, CE
Temperature RangeAmbient to 95°C
DisplayMonochrome LCD
Heating Capacity300 w
IncludesPolycarbonate Gable Cover, Diffuser Tray
MaterialEpoxy Powder-coated Steel
ModelIsotemp GPD 05
Temperature Presets3
Temperature Stability±0.2°C
Temperature Uniformity±0.2°C
Dimensions (D x W x H) Interior6.1 x 11.8 x 5.99 in. (154 x 300 x 150 mm)
Dimensions (L x W x H)9.7 x 14 x 9.1 in. (246 x 355 x 232 mm)
Amperage2.2-2.7 / 1.0-1.3 (120V/230V)
Unit SizeEA