Isotemp™ Heated Bath Circulators: Model 6200 H24
Isotemp™ Heated Bath Circulators: Model 6200 H24
Actual product may vary
Isotemp™ Heated Bath Circulators: Model 6200 H24
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Have Questions?
Catalog NumberFrequency
FTN21131310050 Hz
Catalog number FTN211313100
Price (SGD)
12,667.00
EA
Add to cart
Frequency:
50 Hz
Price (SGD)
12,667.00
EA
Add to cart
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters
  • High temperature and low level cut-outs
  • Compact footprint maximizes bench space
  • Includes external circulation plumbing with 8-mm and 12-mm fittings and supply and return clamps
  • Work area cover included on all heated baths
  • High quality stainless steel baths
  • All bath drains are located on the front of the unit with an integrated valve
Specifications
Pump TypeForce and Suction
Temperature Range10°C to 200°C
Heating Capacity2000 w
Model6200 H24
Dimensions (L x W x H)56.13 x 36.32 x 414 cm
Weight (Metric)16.8 kg
Electrical Requirements230 V 50 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA