Isotemp™ Heated Bath Circulators: Model 6200 H7
Isotemp™ Heated Bath Circulators: Model 6200 H7
Actual product may vary
Isotemp™ Heated Bath Circulators: Model 6200 H7
Isotemp™ Heated Bath Circulators: Model 6200 H7
Isotemp™ Heated Bath Circulators: Model 6200 H7
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Have Questions?
Catalog NumberVoltageFrequency
FTN211311100230 V50 Hz
Catalog number FTN211311100
Price (SGD)
10,636.00
EA
Add to cart
Voltage:
230 V
Frequency:
50 Hz
Price (SGD)
10,636.00
EA
Add to cart
Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters.
  • High temperature and low level cut-outs
  • Compact footprint maximizes bench space
Specifications
Capacity (Metric)5.1 to 6.5 L
Pump TypeForce and Suction
Temperature Range10°C to 200°C
Length (Metric)363.2 mm
Width (Metric)215.9 mm
Height (Metric)414 mm
Weight (Metric)10.9 kg
Voltage230 V
Frequency50 Hz
Unit SizeEA