Isotemp™ 4100 H5P Open Heated PPO Bath Circulators
Isotemp™ 4100 H5P Open Heated PPO Bath Circulators
Actual product may vary
Isotemp™ 4100 H5P Open Heated PPO Bath Circulators
Open heated baths are made from very rigid polyphenyleneoxide (PPO) and are thermally resistant up to +100°C, making them an excellent alternative to stainless steel.
Have Questions?
Catalog NumberElectrical RequirementsModelCapacity (Metric)
FTN211114100230 V 60 Hz4100 H5P4.5 to 5.3 L
Catalog number FTN211114100
Price (SGD)
4,005.00
EA
Add to cart
Electrical Requirements:
230 V 60 Hz
Model:
4100 H5P
Capacity (Metric):
4.5 to 5.3 L
Price (SGD)
4,005.00
EA
Add to cart
Open heated baths are made from very rigid polyphenyleneoxide (PPO) and are thermally resistant up to +100°C, making them an excellent alternative to stainless steel.
These economical open-bath circulators use water as the heat transfer liquid. Baths have integrated grips and supports and are fitted with bridge plates for placement of the control head.
  • Heating capacity up to 1000 watts
  • With 5-liter bath
  • The most economical choice for a heating bath
Temperature range: Ambient to +100°C
Specifications
Capacity (English)1.19 to 1.4 gal.
Capacity (Metric)4.5 to 5.3 L
Flow Rate17 L/min.
Pump TypeForce
Temperature RangeAmbient +10° to 100°C
TypeIsotemp™ Open Heated Bath Circulator
Heating Capacity1.2 kW
MaterialPolyphenyleneoxide
Model4100 H5P
Temperature Stability±0.5°C
Dimensions (L x W x H)15.3 x 7.5 x 13.7 in. (38.8 x 19 x 34.7 cm)
Length (English)15.3 in.
Length (Metric)38.8 cm
Width (English)7.5 in.
Width (Metric)19 cm
Height (English)13.7 in.
Height (Metric)34.7 cm
Weight (English)11.9 lb.
Weight (Metric)5.4 kg
Electrical Requirements230 V 60 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA