accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Actual product may vary
Accumet
accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
Have Questions?
Catalog NumberTypeProbe TypeMaterial
FSP13-636-AB315BpH/mV Meter with ProbeRefillable pH ElectrodeGlass Body
Catalog number FSP13-636-AB315B
Price (SGD)
1,735.00
Each
Type:
pH/mV Meter with Probe
Probe Type:
Refillable pH Electrode
Material:
Glass Body
Price (SGD)
1,735.00
Each
  • Large, backlit display simultaneous shows pH, mV and temperature readings
  • One to five point pH calibration with automatic buffer recognition of four pH buffer sets
  • One point relative mV calibration (RmV) for ORP measurements
  • One point ATC temperature probe calibration
  • Calibration due alarm, selectable 1 to 60 hr., or off
  • Automatic or manual temperature compensation
  • Ready icon indicates measurement stability
  • Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
  • Non-volatile meter memory preserves settings and data in event of power loss
  • Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications
Accuracy (Temperature)±0.3°C (±0.5°F)
Accuracy (mV)±0.2 or ±0.05%, Whichever is Greater
Accuracy (pH)±0.01
Data Management DetailsPrinted or CSV Format
DescriptionpH/mV Meter with pH Glass-body Double Junction Refillable Electrode
OutputsPort for Computer or Printer
IncludespH Electrode, ATC Probe, Electrode Stand, Power Adapter
Measurement ModespH with mV and Temperature, Relative mV with mV and Temperature
CalibrationpH, Relative mV, Temperature
Range (pH)-2 to 18
Resolution (pH)0.01, 0.1
Calibration Points (pH)USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
Range (mV)±2000 mV
Resolution (mV)1 mV, 0.1 RmV
Calibration Points1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
Temperature Range (Metric)-5°C to +105°C
Temperature Range (English)23°F to 221°F
Resolution (Temperature)0.1°C (0.1°F)
Display Type5 in. LCD
Backlight OptionYes
MemoryNon-volatile
Data Points500
Log Function TypeAutomatic, Manual
InputsBNC, ATC
Certifications/ComplianceCE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate
IP RatingIP54
AC AdapterUniversal Power Adapter
Warranty3 Years
MaterialGlass Body
ModelAB315
Probe TypeRefillable pH Electrode
Range (Relative mV)±2000 mV
StabilityIndicator Icons
Temperature CompensationAutomatic, Manual
Channels1
TypepH/mV Meter with Probe
Voltage100/240 V