accumet™ AB250 Benchtop pH/ISE Meters
Actual product may vary
accumet™ AB250 Benchtop pH/ISE Meters
accumet™ AB250 Benchtop pH/ISE Meters
accumet™ AB250 Benchtop pH/ISE Meters
accumet™ AB250 Benchtop pH/ISE Meters
Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB250 pH/ISE meter provides flexible measurements of pH or ion concentration using an ion selective electrode (ISE) in one easy to use, reliable measurement system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
FSP13-636-AB250Meter with pH/ISE Kit
FSP13-636-AB250AAB250 pH/ISE Meter
13-636-AB250Meter with pH/ISE Kit
Catalog number FSP13-636-AB250
Price (SGD)
1,385.00
Each
Product Type:
Meter with pH/ISE Kit
Price (SGD)
1,385.00
Each
Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB250 pH/ISE meter provides flexible measurements of pH or ion concentration using an ion selective electrode (ISE) in one easy to use,
  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (13-620-BSP) with five speed settings
NIST-Traceable Calibration Model (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate
Specifications
  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion), 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Specifications
OutputsRS232 (phono plug), mini-B USB, stirrer
Accuracy (ISE)0.5% full scale (monovalent ion), 1% full scale (divalent ion)
Accuracy (mV)±0.2 mV
Accuracy (pH)±0.002
Calibration Points (ISE)2 to 6 points from one of following group, 0.001, 0.01, 0.1, 1, 10, 100, 0.01, 0.1, 1, 10, 100, 1000
Calibration Points (pH)Up to 6 preset or 5 custom
Certifications/ComplianceWithout NIST-traceable pH Certificate
Data Management DetailsManual, timed (selectable every 3 to 3600 sec.), printer or CSV format
Data Points500
Display TypeLCD
Measurement ModespH, mV, ISE, Ion, Concentration, Temperature
Range (mV)±2000 mV
Range (pH)-2 to 20
Resolution (ISE)0.001/0.01/0.1/1 (automatic)
Resolution (pH)0.1/0.01/0.001
Temperature Range (Metric)0°C to 100°C
IncludesMeter, TRIS Compatible pH/ATC Electrode (13-620-631), Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply
InputsBNC, ATC
ModelAB250 pH/ISE Kit
Product TypeMeter with pH/ISE Kit
Range (ISE)0.001 to 19999 (±2000)
Resolution (mV)0.1mV
Temperature Accuracy±0.3°C
Temperature Resolution0.1°C
Warranty3 Years
Voltage100/240 V
Frequency50/60Hz
Unit SizeEach