Check Order Status
Buffer pH 12.45
Buffer pH 12.45
Buffer pH 12.45 colourless 480 mL ECBU12BT
Have Questions?
Catalog Number
Quantity
FSP01X211212
480 mL
Catalog number
FSP01X211212
Price (SGD)
-
Quantity:
480 mL
Buffer pH 12.45 colourless 480 mL ECBU12BT
Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Color
Colorless
pH
12.45
Quantity
480 mL