▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Buffer Solution pH 4.01
Buffer Solution pH 4.01
Buffer solution pH 4.01 red 480mL ECBU4BTC
Have Questions?
Catalog Number
Quantity
FSP01X211204
480 mL
Catalog number
FSP01X211204
Price (SGD)
25.00
Special offer
Ends:
06-Sep-2026
Each
Add to cart
Quantity:
480 mL
Price (SGD)
25.00
Special offer
Ends:
06-Sep-2026
Each
Add to cart
Buffer solution pH 4.01 red 480mL ECBU4BTC
Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Color
Red
pH
4.01
Quantity
480 mL