Sodium hypochlorite, 11-15% available chlorine
Sodium hypochlorite, 11-15% available chlorine
Sodium hypochlorite, 11-15% available chlorine
Thermo Scientific Chemicals
Sodium hypochlorite, 11-15% available chlorine
CAS: 7681-52-9 | NaOCl | 74.44 g/mol
Have Questions?
Catalog NumberQuantity
FSKL14709.AP500 mL
Catalog number FSKL14709.AP
Price (SGD)
76.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
76.00
Each
Chemical Identifiers
CAS7681-52-9, 7732-18-5, 1310-73-2
Specifications
Boiling Point111°C
Chemical Name or MaterialSodium hypochlorite
Melting Point-16°C
CAS7681-52-9
ColorGreen-Yellow