Sodium hypochlorite, 11-15% available chlorine
Sodium hypochlorite, 11-15% available chlorine
Thermo Scientific Chemicals
Sodium hypochlorite, 11-15% available chlorine
CAS: 7681-52-9 | ClNaO | 74.439 g/mol
Have Questions?
Catalog NumberQuantity
FSKL14709.0F2.5 L
Catalog number FSKL14709.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium Hypochlorite aq. soln.
Name Note11-15% available chlorine
CAS7681-52-9
ColorYellow
Health Hazard 1H290-H314-H335
View more
Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.