Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
Thermo Scientific Chemicals
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
CAS: 555-60-2 | C9H5ClN4 | 204.617 g/mol
Have Questions?
Catalog NumberQuantity
FSKL06932.MC100 mg
Catalog number FSKL06932.MC
Price (SGD)
86.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
86.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbonyl cyanide 3-chlorophenylhydrazone
CAS555-60-2
Health Hazard 1H301+H311+H331-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
View more
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.
Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
  1. A.J.Verhoeven; B.P.Neve; H.Jansen. Secretion and apparent activation of human hepatic lipase requires proper oligosaccharide processing in the endoplasmic reticulum. Biochemical Journal. 1999, 337,(Pt 1), 133-140.
  2. G.Biswas; O.A.Adebanjo; B.D.Freedman; H.K.Anandatheerthavarada; C.Vijayasarathy; M.Zaidi; M.Kotlikoff; N.G.Avadhani. Retrograde Ca2+ signaling in C2C12 skeletal myocytes in response to mitochondrial genetic and metabolic stress: a novel mode of inter-organelle crosstalk. EMBO Journal. 1999, 18,(3), 522-533.
  3. Uncoupler of oxidative phosphorylation: Biochem. Biophys. Acta, 394 145 (1975); Biochemistry, 17, 2245 (1978).