TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
Thermo Scientific Chemicals
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade
Use directly to re-suspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSKJ75793.AE100 mLNalgene bottle
Catalog number FSKJ75793.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTE Buffer
CAS1185-53-1
Recommended StorageAmbient temperatures
Concentration1X
TSCAYes
View more
Use directly to re-suspend and/or dilute purified DNA or RNA.