▸
Water
▸
Life Science Water
▸
Molecular Biology Grade Life Science Water
▸
Water, Nuclease-free, Molecular Biology Grade, Ultrapure
Element Search
Structure Search
Thermo Scientific Chemicals
Water, Nuclease-free, Molecular Biology Grade, Ultrapure
CAS: 7732-18-5 | H
2
O | 18.015 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FSKJ71786.AP
500 mL
Nalgene Bottle
Catalog number
FSKJ71786.AP
Price (SGD)
131.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Nalgene Bottle
Request bulk or custom format
Price (SGD)
131.00
Each
Add to cart
Specifications
Chemical Name or Material
Water
CAS
7732-18-5
Melting Point
0°C
Boiling Point
100°C
Recommended Storage
Ambient temperatures