Formamide, Ultra Pure, 99.5% min
Formamide, Ultra Pure, 99.5% min
Formamide, Ultra Pure, 99.5% min
Formamide, Ultra Pure, 99.5% min
Thermo Scientific Chemicals
Formamide, Ultra Pure, 99.5% min
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ67206.AE100 mL
Catalog number FSKJ67206.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialFormamide
Melting Point2°C
Name NoteUltra Pure
CAS75-12-7
ColorColorless
View more
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.
Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.
Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
RUO – Research Use Only
Ronald. Breslow,; Tao. Guo.Diels-Alder reactions in nonaqueous polar solvents. Kinetic effects of chaotropic and antichaotropic agents and of .beta.-cyclodextrin . J. Am. Chem. Soc. 1988, 110 (17), 5613-5617.