Spectinomycin sulfate, 93%
Spectinomycin sulfate, 93%
Spectinomycin sulfate, 93%
Spectinomycin sulfate, 93%
Thermo Scientific Chemicals
Spectinomycin sulfate, 93%
CAS: 23312-56-3 | C14H26N2O11S | 430.43 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSKJ66034.065 gPlastic bottle
Catalog number FSKJ66034.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSpectinomycin sulfate
CAS23312-56-3
Health Hazard 1H317-H334
Health Hazard 3P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
Recommended StorageKeep cold
View more
Targets Gram-negative bacteria