Ampicillin sodium salt
Ampicillin sodium salt
Ampicillin sodium salt
Ampicillin sodium salt
Thermo Scientific Chemicals
Ampicillin sodium salt
Ampicillin sodium salt, CAS # 69-52-3, is the sodium salt form of ampicillin, a semisynthetic derivative of aminopenicillin compound that shows antibacterial activity against both gram-positive and gram-negative bacteria. | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Catalog number FSKJ63807.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmpicillin sodium salt
TypeAmpicillin Sodium Salt
Physical FormPowder
CAS69-52-3
Health Hazard 1H317-H334
View more
Ampicillin sodium salt is a reagent for transformed cells expressing beta-lactamase. It acts as a structural analogue of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme and prevents the cross-linking of the peptidoglycan of the cell wall necessary for the growth of the bacterium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Ampicillin sodium salt is a semi-synthetic derivative of penicillin with a broad-spectrum antibacterial activity against several species, including Bacillus subtilis, Staphylococcus aureus, Escherichia coli, and Ralstonia solanacearum
• It can inhibit bacterial cell wall synthesis by binding to penicillin-binding proteins, inhibiting peptidoglycan synthesis, which is an essential component of the bacterial cell wall
Application
• Ampicillin sodium salt has been used as a reagent for transformed cells that express β-lactamase
• It can be used to select for ampicillin resistance in mutated and transformed cells
• Ampicillin sodium salt acts as a structural analog of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme preventing the cross-linking of the cell wall peptidoglycan, which is necessary for the growth of the bacterium
RUO – Research Use Only
General References:
  1. Hou, X.; Hansen, J. H.; Bjerre, A. B. Integrated bioethanol and protein production from brown seaweed Laminaria digitata. Bioresour. Technol. 2015, 197, 310-317.
  2. Tang, L.; Su, M.; Yan, J.; Xie, S.; Zhang, W. Lid hinge region of Penicillium expansum lipase affects enzyme activity and interfacial activation. Process Biochem. 2015, 50 (8), 1218-1223.