myo-Inositol, Cell Culture Grade
myo-Inositol, Cell Culture Grade
myo-Inositol, Cell Culture Grade
myo-Inositol, Cell Culture Grade
Thermo Scientific Chemicals
myo-Inositol, Cell Culture Grade
A lipotropic agent | CAS: 87-89-8 | C6H12O6 | 180.156 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ62886.1850 g
Catalog number FSKJ62886.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Materialmyo-Inositol
CAS87-89-8
Melting Point222°C to 227°C
ColorWhite
Recommended StorageAmbient temperatures
View more
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
Solubility
It is soluble in water (50 mg/ml).
Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
  1. Robert B Campenot et. al. Production of compartmented cultures of rat sympathetic neurons. Nature Protocols. 20094 (12), 1869-1887.
  2. M A Ferguson and A F Williams. Cell-surface anchoring of proteins via glycosyl-phosphatidylinositol structures. Biochemistry. 198857 (1), 285-320.