Glycine, 0.2M buffer soln., pH 3.0
Glycine, 0.2M buffer soln., pH 3.0
Glycine, 0.2M buffer soln., pH 3.0
Glycine, 0.2M buffer soln., pH 3.0
Thermo Scientific Chemicals
Glycine, 0.2M buffer soln., pH 3.0
CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ62527.AK250 mL
Catalog number FSKJ62527.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycine
CAS56-40-6
Recommended StorageAmbient temperatures
Concentration0.2 M
pH3.0
View more
Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycine is widely used as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
Solubility
Not miscible or difficult to mix in water. Slightly soluble in acetone. Insoluble in alcohol, and ether.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only
General References:
  1. H Towbin, et al. Electrophoretic transfer of proteins from polyacrylamide gels to nitrocellulose sheets: procedure and some applications.Proc Natl Acad Sci U S A.,1979,76(9), 4350-4354.
  2. Laemmli UK, et al. Cleavage of structural proteins during the assembly of the head of bacteriophage T4.Nature.,1970,227(5259), 680-5.