Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Thermo Scientific Chemicals
Formaldehyde, 4% in PBS
Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ60401.AK250 mL
Catalog number FSKJ60401.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialFormaldehyde
Physical FormLiquid
CAS50-00-0
Health Hazard 1H315-H317-H319-H341-H350
Health Hazard 3P201-P202-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P501c
View more
Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.