Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade
Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade
Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade
Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade
Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade
CAS: 51805-45-9 | C9H12O6P | 247.16 g/mol
Have Questions?
Catalog NumberQuantity
FSKJ60316.0910 g
Catalog number FSKJ60316.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTris(2-carboxyethyl)phosphine hydrochloride
CAS51805-45-9
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
Melting Point177°C to 178°C
View more
For reducing disulfide bonds in peptides and proteins.