▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
Tween 20™, Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
Tween 20™, Ultrapure, Thermo Scientific Chemicals
Have Questions?
Catalog Number
Quantity
Packaging
FSKJ20605.AP
500 mL
Plastic bottle
Catalog number
FSKJ20605.AP
Price (SGD)
-
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Tween 20™
CAS
9005-64-5
Physical Form
Liquid
Boiling Point
>100°C
Recommended Storage
Ambient temperatures