Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
Chelating agent used in many enzyme buffers and at higher concentrations, as enzyme inactivator | CAS: 6381-92-6 | C10H18N2Na2O10 | 372.238 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSKJ15694.AE100 mLNalgene bottle
Catalog number FSKJ15694.AE
Price (SGD)
107.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (SGD)
107.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid
CAS6381-92-6
Health Hazard 1H373
Health Hazard 3P260-P314-P501c
Recommended StorageAmbient temperatures
View more
Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.