2-Phenylacetamide, 99%
2-Phenylacetamide, 99%
2-Phenylacetamide, 99%
Thermo Scientific Chemicals
2-Phenylacetamide, 99%
CAS: 103-81-1 | C8H9NO | 135.17 g/mol
Have Questions?
Catalog NumberQuantity
FSKH60626.MD250 mg
Catalog number FSKH60626.MD
Price (SGD)
118.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (SGD)
118.00
Each
Specifications
Chemical Name or Material2-Phenylacetamide
CAS103-81-1
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store at 4°C.
RUO – Research Use Only