Quinaldine Red, dye content, 95%
Quinaldine Red, dye content, 95%
Quinaldine Red, dye content, 95%
Quinaldine Red, dye content, 95%
Thermo Scientific Chemicals
Quinaldine Red, dye content, 95%
CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol
Have Questions?
Catalog NumberQuantity
FSKH55729.065 g
Catalog number FSKH55729.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialQuinaldine Red
Name NoteDye content
CAS117-92-0
ColorBlack to Green
Recommended StorageAmbient temperatures
View more
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).
Solubility
Partly soluble in water, alcohol, and ethanol.
Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Elizabeth B. Cogan.; G.Bruce Birrell.; O.Hayes Griffith. A Robotics-Based Automated Assay for Inorganic and Organic Phosphates. Anal. Biochem. 1999, 271 (1), 29-35.
  2. Hitoshi Imamura.; Toru Maruyama.; Hiroaki Okabe.; Hidekata Shimada.; Masaki Otagiri. A Simple and Rapid Fluorometric Determination Method of α1-Acid Glycoprotein in Serum Using Quinaldine Red.Pharm. Res.1994, 11 (4),566-570.