2-Amino-5-bromopyrazine, 97%
2-Amino-5-bromopyrazine, 97%
2-Amino-5-bromopyrazine, 97%
2-Amino-5-bromopyrazine, 97%
Thermo Scientific Chemicals
2-Amino-5-bromopyrazine, 97%
CAS: 59489-71-3 | C4H4BrN3 | 174.001 g/mol
Have Questions?
Catalog NumberQuantity
FSKH27465.065 g
Catalog number FSKH27465.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-bromopyrazine
CAS59489-71-3
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c